LY-3200882 98%

Warning

Synonyms: 2-(4-((4-((1-Cyclopropyl-3-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-yl)oxy)pyridin-2-yl)amino)pyridin-2-yl)propan-2-ol, 4-[[4-[[1-cyclopropyl-3-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-yl]oxy]-2-pyridinyl]amino]-α,α-dimethyl-2-pyridinemethanol, LY3200882

A352832-10MG A352832-1MG A352832-25MG A352832-5MG
76648-500 76648-502 76648-504 76648-506
LY-3200882 98%
LY-3200882

Precaución: for Research Use Only

Formula: C₂₄H₂₉N₅O₃
MW: 435.53 g/mol
MDL Number: MFCD31619238
CAS Number: 1898283-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR