Trifluoromethyl 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

A352130-100MG A352130-1G A352130-250MG A352130-5G A352130-25G
76648-492EA 30.77 USD
76648-492 76648-494 76648-496 76648-498 77748-691
Trifluoromethyl 4-methylbenzenesulfonate 95%
Trifluoromethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₈H₇F₃O₃S
MW: 240.2 g/mol
MDL Number: MFCD31695793
CAS Number: 175676-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR