(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S,R,S)-AHPC-Me
A351331-100MG
A351331-1G
A351331-1MG
A351331-250MG
A351331-50MG
A351331-5G
76648-482EA
47.75
USD
76648-482
76648-484
76648-486
76648-488
76648-490
77572-032
(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide 97%
(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide
Precaución: for Research Use Only
Formula:
C₂₃H₃₂N₄O₃S MW: 444.59 g/mol |
MDL Number:
MFCD31630815 CAS Number: 1948273-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...