tert-Butyl 2-bromo-1H-imidazole-1-carboxylate 95%

Supplier: Ambeed
Warning

A351074-100MG A351074-1G A351074-250MG A351074-5G A351074-25G
76648-474EA 26.53 USD
76648-474 76648-476 76648-478 76648-480 77748-690
tert-Butyl 2-bromo-1H-imidazole-1-carboxylate 95%
tert-Butyl 2-bromo-1H-imidazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O₂
MW: 247.0891 g/mol
MDL Number: MFCD30189396
CAS Number: 1207457-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR