Porcupine-IN-1 97%

Supplier: Ambeed
Warning

Synonyms: Porcn-IN-1, N-((3-Fluoro-2'-methyl-[2,4'-bipyridin]-5-yl)methyl)-9H-carbazole-2-carboxamide

A349677-10MG A349677-1MG A349677-25MG A349677-50MG A349677-5MG
76648-464EA 71.09 USD
76648-464 76648-466 76648-468 76648-470 76648-472
Porcupine-IN-1 97%
Porcupine-IN-1

Precaución: for Research Use Only

Formula: C₂₅H₁₉FN₄O
MW: 410.44 g/mol
MDL Number: MFCD31630804
CAS Number: 2036044-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR