1-(3-Chloro-5-methylpyridin-4-yl)ethan-1-one 98%

Supplier: Ambeed
Warning

A1340760-100MG A1340760-250MG
76658-066EA 359.65 USD
76658-066 76658-068
1-(3-Chloro-5-methylpyridin-4-yl)ethan-1-one 98%
1-(3-Chloro-5-methylpyridin-4-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.6082 g/mol
MDL Number: MFCD18258082
CAS Number: 1256809-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR