(S)-2-Amino-3-guanidinopropanoic acid dihydrochloride 95%

Supplier: Ambeed
Warning

A346871-1G A346871-250MG A346871-5G
76648-448EA 88 USD
76648-448 76648-450 76648-452
(S)-2-Amino-3-guanidinopropanoic acid dihydrochloride 95%
(S)-2-Amino-3-guanidinopropanoic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂Cl₂N₄O₂
CAS Number: 2216751-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR