(2S,4R)-1-((S)-2-(3-(2-((5-((4-((3-chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)pentyl)oxy)ethoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide ≥99%

Supplier: Ambeed
Warning

Synonyms: Gefitinib-based PROTAC 3

A346407-10MG A346407-1MG A346407-25MG A346407-50MG A346407-5MG
76648-438EA 61.54 USD
76648-438 76648-440 76648-442 76648-444 76648-446
(2S,4R)-1-((S)-2-(3-(2-((5-((4-((3-chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)pentyl)oxy)ethoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide ≥99%
(2S,4R)-1-((S)-2-(3-(2-((5-((4-((3-chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)pentyl)oxy)ethoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₄₇H₅₇ClFN₇O₈S
MW: 934.51 g/mol
MDL Number: MFCD32062811
CAS Number: 2230821-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR