Methyl 2-(2-(2-chloro-5-((R)-2-hydroxy-3-(methylamino)propoxy)phenyl)-6-(3,5-dimethylisoxazol-4-yl)-5-methylpyrimidin-4-yl)-2,7-diazaspiro[3.5]nonane-7-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: EZM 2302

A345312-1MG A345312-5MG
76648-428EA 59 USD
76648-428 76648-430
Methyl 2-(2-(2-chloro-5-((R)-2-hydroxy-3-(methylamino)propoxy)phenyl)-6-(3,5-dimethylisoxazol-4-yl)-5-methylpyrimidin-4-yl)-2,7-diazaspiro[3.5]nonane-7-carboxylate 97%
Methyl 2-(2-(2-chloro-5-((R)-2-hydroxy-3-(methylamino)propoxy)phenyl)-6-(3,5-dimethylisoxazol-4-yl)-5-methylpyrimidin-4-yl)-2,7-diazaspiro[3.5]nonane-7-carboxylate

Precaución: for Research Use Only

Formula: C₂₉H₃₇ClN₆O₅
MDL Number: MFCD31657380
CAS Number: 1628830-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR