6'-Amino-N-[2-(morpholin-4-yl)-[1,3]oxazolo[4,5-b]pyridin-6-yl]-[2,3'-bipyridine]-6-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: IRAK4-IN-7

A345196-10MG A345196-1MG A345196-2MG A345196-50MG A345196-5MG A345196-100MG
76648-418EA 65.78 USD
76648-418 76648-420 76648-422 76648-424 76648-426 77748-678
6'-Amino-N-[2-(morpholin-4-yl)-[1,3]oxazolo[4,5-b]pyridin-6-yl]-[2,3'-bipyridine]-6-carboxamide 98%
6'-Amino-N-[2-(morpholin-4-yl)-[1,3]oxazolo[4,5-b]pyridin-6-yl]-[2,3'-bipyridine]-6-carboxamide

Precaución: for Research Use Only

Formula: C₂₁H₁₉N₇O₃
MW: 417.42 g/mol
MDL Number: MFCD31619276
CAS Number: 1801343-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR