N-(3-(2-(2-Hydroxyethoxy)-6-morpholinopyridin-4-yl)-4-methylphenyl)-2-(trifluoromethyl)isonicotinamide 97%

Supplier: Ambeed
Warning

Synonyms: LXH254

A344788-1MG A344788-25MG A344788-5MG A344788-100MG A344788-250MG A344788-50MG A344788-10MG
76648-412EA 31.83 USD
76648-412 76648-414 76648-416 77598-430 77598-432 77598-434 77616-512
N-(3-(2-(2-Hydroxyethoxy)-6-morpholinopyridin-4-yl)-4-methylphenyl)-2-(trifluoromethyl)isonicotinamide 97%
N-(3-(2-(2-Hydroxyethoxy)-6-morpholinopyridin-4-yl)-4-methylphenyl)-2-(trifluoromethyl)isonicotinamide

Precaución: for Research Use Only

Formula: C₂₅H₂₅F₃N₄O₄
MW: 502.49 g/mol
MDL Number: MFCD31657408
CAS Number: 1800398-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR