3-Amino-2-chloropyridine 98+%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

A343231-100G A343231-25G A343231-500G A343231-1000G
76648-406EA 22 USD
76648-406 76648-408 76648-410 77622-284
3-Amino-2-chloropyridine 98+%
3-Amino-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 263
MDL Number: MFCD00006238
CAS Number: 6298-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR