5-Methylpyridin-2(1H)-one 98%

Supplier: Ambeed
Danger

A342907-25G A342907-5G A342907-100G
76648-400EA 10.61 USD
76648-400 76648-402 76648-404 77616-504 77748-671
5-Methylpyridin-2(1H)-one 98%
5-Methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₇NO
MDL Number: MFCD00955788
CAS Number: 1003-68-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR