2-Chloro-1-ethynyl-4-nitrobenzene 95%

Supplier: Ambeed
Warning

A339997-100MG A339997-1G A339997-250MG
76648-384EA 27 USD
76648-384 76648-386 76648-388
2-Chloro-1-ethynyl-4-nitrobenzene 95%
2-Chloro-1-ethynyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂
MW: 181.5759 g/mol
MDL Number: MFCD11036434
CAS Number: 1260829-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR