2-(Difluoromethyl)-5-iodopyridine 95%

Supplier: Ambeed
Warning

A339125-100MG A339125-1G A339125-250MG
76648-376EA 16 USD
76648-376 76648-378 76648-380
2-(Difluoromethyl)-5-iodopyridine 95%
2-(Difluoromethyl)-5-iodopyridine

Precaución: for Research Use Only

Formula: C₆H₄F₂IN
MDL Number: MFCD25476549
CAS Number: 1805287-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR