(S)-tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethyl)carbamate 95%

Supplier: Ambeed
Warning

A339064-100MG A339064-1G A339064-250MG A339064-5G
76648-370EA 59.42 USD
76648-370 76648-372 76648-374 77748-668
(S)-tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethyl)carbamate 95%
(S)-tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₃₀BNO₄
MW: 347.2568 g/mol
MDL Number: MFCD29912577
CAS Number: 1171897-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR