4-(5-(3,5-Dichlorophenyl)-5-(trifluoromethyl)-4,5-dihydroisoxazol-3-yl)-N-((methoxyamino)methylene)-2-methylbenzamide ≥99%
Supplier: Ambeed
|
Warning
|
A334893-10MG
A334893-25MG
A334893-5MG
A334893-1MG
76648-364EA
141
USD
76648-364
76648-366
76648-368
77598-428
4-(5-(3,5-Dichlorophenyl)-5-(trifluoromethyl)-4,5-dihydroisoxazol-3-yl)-N-((methoxyamino)methylene)-2-methylbenzamide ≥99%
4-(5-(3,5-Dichlorophenyl)-5-(trifluoromethyl)-4,5-dihydroisoxazol-3-yl)-N-((methoxyamino)methylene)-2-methylbenzamide
Precaución: for Research Use Only
Formula:
C₂₀H₁₆Cl₂F₃N₃O₃ |
MDL Number:
MFCD31692349 CAS Number: 928783-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...