Benzo[d][1,3]dioxol-5-yl carbonochloridate 95%

Supplier: Ambeed
Danger

A334477-1G A334477-250MG A334477-5G A334477-100MG
76648-358EA 190.97 USD
76648-358 77748-660 77748-661 77748-659
Benzo[d][1,3]dioxol-5-yl carbonochloridate 95%
Benzo[d][1,3]dioxol-5-yl carbonochloridate

Precaución: for Research Use Only

Formula: C₈H₅ClO₄
MW: 200.5800 g/mol
CAS Number: 117886-89-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR