Nitd-349 97%

Supplier: Ambeed
Warning

Synonyms: N-(4,4-Dimethylcyclohexyl)-4,6-difluoro-1H-indole-2-carboxamide

A334115-10MG A334115-1MG A334115-25MG A334115-5MG
76648-350EA 98.67 USD
76648-350 76648-352 76648-354 76648-356
Nitd-349 97%
Nitd-349

Precaución: for Research Use Only

Formula: C₁₇H₂₀F₂N₂O
MW: 306.35 g/mol
MDL Number: MFCD31630740
CAS Number: 1473450-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR