4-(Dichloromethyl)benzonitrile 95%

Supplier: Ambeed
Warning

A333068-100MG A333068-10G A333068-1G A333068-250MG
76648-338EA 48 USD
76648-338 76648-340 76648-342 76648-344
4-(Dichloromethyl)benzonitrile 95%
4-(Dichloromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MDL Number: MFCD00045606
CAS Number: 74231-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR