(S)-tert-Butyl 3-(cyanomethyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A332826-1G A332826-5G A332826-100MG A332826-250MG
76648-334EA 546.37 USD
76648-334 76648-336 77593-962 77593-964
(S)-tert-Butyl 3-(cyanomethyl)piperazine-1-carboxylate 97%
(S)-tert-Butyl 3-(cyanomethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉N₃O₂
MW: 225.2875 g/mol
MDL Number: MFCD31978023
CAS Number: 1589082-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR