VPS34 inhibitor 1 95%

Supplier: Ambeed
Warning

A332678-10MG A332678-1MG A332678-25MG A332678-5MG
76648-326EA 338 USD
76648-326 76648-328 76648-330 76648-332
VPS34 inhibitor 1 95%
VPS34 inhibitor 1

Precaución: for Research Use Only

Formula: C₂₁H₂₅N₇O
CAS Number: 1383716-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR