(6S,9S,9aS)-N-Benzyl-6-(4-hydroxybenzyl)-2,9-dimethyl-4,7-dioxo-8-(quinolin-8-ylmethyl)hexahydro-2H-pyrazino[2,1-c][1,2,4]triazine-1(6H)-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: C-82

A329844-10MG A329844-1MG A329844-25MG A329844-5MG
76648-312EA 440.28 USD
76648-312 76648-314 76648-316 76648-318
(6S,9S,9aS)-N-Benzyl-6-(4-hydroxybenzyl)-2,9-dimethyl-4,7-dioxo-8-(quinolin-8-ylmethyl)hexahydro-2H-pyrazino[2,1-c][1,2,4]triazine-1(6H)-carboxamide 97%
(6S,9S,9aS)-N-Benzyl-6-(4-hydroxybenzyl)-2,9-dimethyl-4,7-dioxo-8-(quinolin-8-ylmethyl)hexahydro-2H-pyrazino[2,1-c][1,2,4]triazine-1(6H)-carboxamide

Precaución: for Research Use Only

Formula: C₃₃H₃₄N₆O₄
MW: 578.6609 g/mol
MDL Number: MFCD32664545
CAS Number: 1422253-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR