(6-Methoxypyridin-2-yl)methanamine dihydrochloride 95%

Supplier: Ambeed
Warning

A328616-100MG A328616-1G A328616-250MG A328616-5G
76648-304EA 30.77 USD
76648-304 76648-306 76648-308 76648-310
(6-Methoxypyridin-2-yl)methanamine dihydrochloride 95%
(6-Methoxypyridin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₂N₂O
MW: 211.0890 g/mol
MDL Number: MFCD31619130
CAS Number: 2172852-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR