4-((4-Benzyl-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)thio)pyrido[3',2':4,5]thieno[3,2-d]pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: TH1020

A1340564-10MG A1340564-1MG A1340564-25MG A1340564-50MG A1340564-5MG
76658-054EA 50.93 USD
76658-054 76658-056 76658-058 76658-060 76658-062
4-((4-Benzyl-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)thio)pyrido[3',2':4,5]thieno[3,2-d]pyrimidine 98%
4-((4-Benzyl-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)thio)pyrido[3',2':4,5]thieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₂₃H₁₅N₇S₂
MW: 453.5421 g/mol
MDL Number: MFCD30480930
CAS Number: 1841460-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR