tert-Butyl 3-oxoindoline-1-carboxylate 98%

Supplier: Ambeed
Warning

A325913-100MG A325913-1G A325913-250MG A325913-25G A325913-5G
76648-274EA 16.98 USD
76648-274 76648-276 76648-278 76648-280 76648-282
tert-Butyl 3-oxoindoline-1-carboxylate 98%
tert-Butyl 3-oxoindoline-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
MW: 233.2631 g/mol
CAS Number: 369595-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR