6-Chloro-2-phenyl-1H-indole 95%

Supplier: Ambeed
Warning

A324316-100MG A324316-250MG A324316-1G
76648-268EA 85 USD
76648-268 76648-270 77571-920
6-Chloro-2-phenyl-1H-indole 95%
6-Chloro-2-phenyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClN
MDL Number: MFCD11976328
CAS Number: 57039-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR