(2-Chloroquinolin-7-yl)methanol 95%

Supplier: Ambeed
Danger

A323066-100MG A323066-1G A323066-250MG
76648-248EA 167 USD
76648-248 76648-250 76648-252
(2-Chloroquinolin-7-yl)methanol 95%
(2-Chloroquinolin-7-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO
MDL Number: MFCD30188586
CAS Number: 1823116-42-2
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR