3,6,9,12,15,18-Hexaoxaicosane-1,20-dioic acid 98%

Supplier: Ambeed
Warning

A322345-1G A322345-250MG A322345-100MG
76648-244EA 183 USD
76648-244 76648-246 77748-632
3,6,9,12,15,18-Hexaoxaicosane-1,20-dioic acid 98%
3,6,9,12,15,18-Hexaoxaicosane-1,20-dioic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₆O₁₀
MDL Number: MFCD30458028
CAS Number: 77855-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR