Methyl 4-methyl-1H-indazole-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A321589-100MG
A321589-1G
A321589-250MG
A321589-50MG
A321589-5G
76648-234EA
157.02
USD
76648-234
76648-236
76648-238
76648-240
76648-242
Methyl 4-methyl-1H-indazole-3-carboxylate 95%
Methyl 4-methyl-1H-indazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀N₂O₂ MW: 190.1986 g/mol |
MDL Number:
MFCD22556712 CAS Number: 1352415-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...