8-Bromo-5-fluoroisoquinoline 95%

Supplier: Ambeed
Warning

A320964-100MG A320964-1G A320964-250MG A320964-5G
76648-226EA 193.09 USD
76648-226 76648-228 76648-230 76648-232
8-Bromo-5-fluoroisoquinoline 95%
8-Bromo-5-fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.0451 g/mol
MDL Number: MFCD11846307
CAS Number: 1378874-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR