Methyl 6-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

A320571-1G A320571-250MG A320571-100MG
76648-222EA 249.32 USD
76648-222 76648-224 77748-626
Methyl 6-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate 95%
Methyl 6-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.0314 g/mol
MDL Number: MFCD30721037
CAS Number: 1807201-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR