4-Chloro-2-ethyl-1-fluorobenzene 95%

Supplier: Ambeed
Warning

A320252-100MG A320252-1G A320252-250MG
76648-216EA 82.76 USD
76648-216 76648-218 76648-220
4-Chloro-2-ethyl-1-fluorobenzene 95%
4-Chloro-2-ethyl-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClF
MW: 158.6005 g/mol
MDL Number: MFCD22490709
CAS Number: 1369889-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR