N-Cbz-piperidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A317760-25G A317760-5G
76648-208EA 45.62 USD
76648-208 76648-210
N-Cbz-piperidine-2-carboxylic acid 98%
N-Cbz-piperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.2891 g/mol
MDL Number: MFCD01863568
CAS Number: 71170-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR