9-Chloro-7-methyl-1,2,3,4-tetrahydroacridine 97%

Supplier: Ambeed
Warning

A1340560-100MG A1340560-1G A1340560-250MG A1340560-5G
76658-046EA 46.68 USD
76658-046 76658-048 76658-050 76658-052
9-Chloro-7-methyl-1,2,3,4-tetrahydroacridine 97%
9-Chloro-7-methyl-1,2,3,4-tetrahydroacridine

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClN
MW: 231.7207 g/mol
MDL Number: MFCD11849269
CAS Number: 5778-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR