Ethyl 2H-1,2,3-triazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A316270-100MG A316270-250MG A316270-1G
76648-200EA 59 USD
76648-200 76648-202 77616-388
Ethyl 2H-1,2,3-triazole-4-carboxylate 95%
Ethyl 2H-1,2,3-triazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MDL Number: MFCD27959308
CAS Number: 1084802-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR