6-Bromoquinoline-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A313175-250MG A313175-1G
76648-186EA 20.16 USD
76648-186 77598-414
6-Bromoquinoline-2,4(1H,3H)-dione 95%
6-Bromoquinoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.0534 g/mol
MDL Number: MFCD29472381
CAS Number: 138964-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR