Hydroxy-PEG6-CH2CO2tBu 98%

Supplier: Ambeed
Warning

Synonyms: Hydroxy-PEG6-CH2-Boc, tert-Butyl 20-hydroxy-3,6,9,12,15,18-hexaoxaicosanoate

A312420-100MG A312420-1G A312420-250MG A312420-5G
76648-180EA 45.62 USD
76648-180 76648-182 76648-184 77748-609
Hydroxy-PEG6-CH2CO2tBu 98%
Hydroxy-PEG6-CH2CO2tBu

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₉
MW: 396.473 g/mol
MDL Number: MFCD27977498
CAS Number: 297162-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR