2,5-Dimethoxypyridine 95%

Supplier: Ambeed
Warning

A311075-100G A311075-1G A311075-250MG A311075-25G A311075-5G
76648-162EA 339 USD
76648-162 76648-164 76648-166 76648-168 76648-170
2,5-Dimethoxypyridine 95%
2,5-Dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD12546620
CAS Number: 867267-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR