2-Methyl-5-nitro-3-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A311034-1G A311034-250MG A311034-5G
76648-156EA 181.42 USD
76648-156 76648-158 76648-160
2-Methyl-5-nitro-3-(trifluoromethyl)pyridine 95%
2-Methyl-5-nitro-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₂
MW: 206.1220 g/mol
MDL Number: MFCD20923732
CAS Number: 1824299-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR