(6-Hydroxyhexyl)triphenylphosphonium bromide 95%
Supplier: Ambeed
|
Warning
|
A310091-100MG
A310091-1G
A310091-250MG
76648-150EA
65
USD
76648-150
76648-152
76648-154
(6-Hydroxyhexyl)triphenylphosphonium bromide 95%
(6-Hydroxyhexyl)triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₂₄H₂₈BrOP MW: 443.3563 g/mol |
MDL Number:
MFCD03265703 CAS Number: 68760-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...