4-Chloro-2-methoxynicotinic acid 95%

Supplier: Ambeed
Warning

A309770-100MG A309770-1G A309770-250MG A309770-5G
76648-142EA 7.43 USD
76648-142 76648-144 76648-146 76648-148
4-Chloro-2-methoxynicotinic acid 95%
4-Chloro-2-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD13188728
CAS Number: 605661-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR