2-(2-Fluorophenoxy)-4-(1-methyl-1H-pyrazol-5-yl)benzamide 98%

Supplier: Ambeed
Warning

A1340414-10MG A1340414-1MG A1340414-25MG A1340414-50MG A1340414-5MG
76658-034EA 125 USD
76658-034 76658-036 76658-038 76658-040 76658-042
2-(2-Fluorophenoxy)-4-(1-methyl-1H-pyrazol-5-yl)benzamide 98%
2-(2-Fluorophenoxy)-4-(1-methyl-1H-pyrazol-5-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₇H₁₄FN₃O₂
MDL Number: MFCD32856863
CAS Number: 2682114-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR