4,6-Dichloro-1-(oxan-2-yl)-1H-pyrazolo[3,4-d]pyrimidine 98%

Supplier: Ambeed
Warning

A306752-1G A306752-250MG A306752-5G A306752-25G
76648-122EA 22 USD
76648-122 76648-124 76648-126 77616-328
4,6-Dichloro-1-(oxan-2-yl)-1H-pyrazolo[3,4-d]pyrimidine 98%
4,6-Dichloro-1-(oxan-2-yl)-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₄O
MW: 273.1186 g/mol
MDL Number: MFCD26394969
CAS Number: 1037479-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR