GSK'872 99+%

Supplier: Ambeed
Warning

Synonyms: N-(6-(Isopropylsulphonyl)quinolin-4-yl)benzo[d]thiazol-5-amine, GSK-872

A306277-10MG A306277-1MG A306277-25MG A306277-5MG A306277-50MG
76648-114EA 54.11 USD
76648-114 76648-116 76648-118 76648-120 77616-322
GSK'872 99+%
GSK'872

Precaución: for Research Use Only

Formula: C₁₉H₁₇N₃O₂S₂
MW: 383.49 g/mol
MDL Number: MFCD30481302
CAS Number: 1346546-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR