Methyl 6-chloro-5-nitropicolinate 95%

Supplier: Ambeed
Warning

A302355-100MG A302355-10G A302355-1G A302355-250MG A302355-5G
76648-094EA 12.74 USD
76648-094 76648-096 76648-098 76648-100 76648-102
Methyl 6-chloro-5-nitropicolinate 95%
Methyl 6-chloro-5-nitropicolinate

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O₄
MW: 216.5786 g/mol
MDL Number: MFCD28139347
CAS Number: 1803583-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR