3-Fluoro-2-methoxyisonicotinic acid 95%

Supplier: Ambeed
Danger

A298834-100MG A298834-1G A298834-250MG A298834-5G
76648-070EA 76.39 USD
76648-070 76648-072 76648-074 76648-076
3-Fluoro-2-methoxyisonicotinic acid 95%
3-Fluoro-2-methoxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MW: 171.1300 g/mol
MDL Number: MFCD13185871
CAS Number: 1214362-24-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR