3''-(tert-Butyl)-4'-(2-hydroxyethoxy)-4''-(pyrrolidin-1-yl)-[1,1':3',1''-terphenyl]-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A298564-1MG A298564-50MG A298564-5MG A298564-100MG A298564-1G A298564-250MG
76648-062EA 10.61 USD
76648-062 76648-064 76648-066 76648-068 77593-808 77593-810 77593-812
3''-(tert-Butyl)-4'-(2-hydroxyethoxy)-4''-(pyrrolidin-1-yl)-[1,1':3',1''-terphenyl]-4-carboxylic acid 97%
3''-(tert-Butyl)-4'-(2-hydroxyethoxy)-4''-(pyrrolidin-1-yl)-[1,1':3',1''-terphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₉H₃₃NO₄
MDL Number: MFCD30481310
CAS Number: 895542-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR