4,4'-Disulfanediylbis(2-methylphenol) 97%

Supplier: Ambeed
Warning

A1340193-1G A1340193-250MG A1340193-5G
76658-020EA 46.68 USD
76658-020 76658-022 76658-024
4,4'-Disulfanediylbis(2-methylphenol) 97%
4,4'-Disulfanediylbis(2-methylphenol)

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂S₂
MW: 278.3898 g/mol
CAS Number: 33931-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR