Calcium chloride 97%

Supplier: Ambeed
Warning

A1340108-100G A1340108-500G
76658-016EA 15 USD
76658-016 76658-018 77566-620
Calcium chloride 97%
Calcium chloride

Precaución: For research use only.

Formula: CaCl₂
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR